missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxypyridine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. M21891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21891G 1 g
M21895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21891G Supplier TCI America Supplier No. M21891G
Only null left

Chemical Identifiers

CAS 26893-73-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD06800962
InChI Key KSWBODXXZITTPO-UHFFFAOYSA-N
Synonym 6-methoxypicolinic acid, 6-methoxy-2-pyridinecarboxylic acid, 2-methoxy-6-picolinic acid, 2-methoxy-6-picolinicacid, 2-pyridinecarboxylic acid, 6-methoxy, picolinic acid, 6-methoxy, 6-methoxy-pyridine-2-carboxylic acid, pubchem15165, 6-methoxypyridine-2-carboxylicacid, acmc-1cccg
PubChem CID 570891
IUPAC Name 6-methoxypyridine-2-carboxylic acid
SMILES COC1=CC=CC(=N1)C(=O)O

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 1 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxypyridine-2-carboxylic Acid