missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxypyridine-2-carboxylic acid, 97%

Catalog No. AAH5965203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5965203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5965203 Supplier Thermo Scientific Chemicals Supplier No. H5965203
Only null left

CAS: 26893-73-2 | C7H7NO3 | 153.137 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with

Chemical Identifiers

CAS 26893-73-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD06800962
InChI Key KSWBODXXZITTPO-UHFFFAOYSA-N
Synonym 6-methoxypicolinic acid, 6-methoxy-2-pyridinecarboxylic acid, 2-methoxy-6-picolinic acid, 2-methoxy-6-picolinicacid, 2-pyridinecarboxylic acid, 6-methoxy, picolinic acid, 6-methoxy, 6-methoxy-pyridine-2-carboxylic acid, pubchem15165, 6-methoxypyridine-2-carboxylicacid, acmc-1cccg
PubChem CID 570891
IUPAC Name 6-methoxypyridine-2-carboxylic acid
SMILES COC1=CC=CC(=N1)C(=O)O

Specifications

Melting Point 130°C to 134°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 6-Methoxypyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only