missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxypyridine-2-carboxaldehyde, 96%

Catalog No. AAH3443203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3443203 1 g
AAH3443206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3443203 Supplier Thermo Scientific Chemicals Supplier No. H3443203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54221-96-4 | C7H7NO2 | 137.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54221-96-4
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD05664030
InChI Key YDNWTNODZDSPNZ-UHFFFAOYSA-N
Synonym 6-methoxypicolinaldehyde, 6-methoxy-2-pyridinecarboxaldehyde, 2-methoxy-6-pyridinecarboxaldehyde, 2-formyl-6-methoxypyridine, 6-methoxy-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-methoxy, 6-methoxypyridine-2-carboxaldehyde, pubchem15345, acmc-1aulb, ksc490s1l
PubChem CID 12325392
IUPAC Name 6-methoxypyridine-2-carbaldehyde
SMILES COC1=CC=CC(=N1)C=O

Specifications

Refractive Index 1.531
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 137.14
Percent Purity 96%
Chemical Name or Material 6-Methoxypyridine-2-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only