missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxypyridine-2-boronic acid pinacol ester, 97%

Catalog No. AAH2726303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2726303 1 g
AAH2726306 5 g
AAH2726314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2726303 Supplier Thermo Scientific Chemicals Supplier No. H2726303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1034297-69-2 | C12H18BNO3 | 235.09 g/mol

It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1034297-69-2
Molecular Formula C12H18BNO3
Molecular Weight (g/mol) 235.09
MDL Number MFCD06798266
InChI Key VYWCRVNLPOVYJH-UHFFFAOYSA-N
Synonym 2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxypyridine-2-boronic acid pinacol ester, pyridine,2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-methoxy pyridine-2-boronic acid pinacol ester, 6-methoxypyridine-2-boronicacidpinacolester, 6-methoxypyridin-2-ylboronic acid pinacol ester, 6-methoxypyridin-2-yl boronic acid pinacol ester, 2-methoxy-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-methoxy-2-pyridinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyloxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 17750508
IUPAC Name 2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)OC

Specifications

Melting Point 68°C to 72°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 235.09
Percent Purity 97%
Chemical Name or Material 6-Methoxypyridine-2-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only