missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyindole 98.0+%, TCI America™

Catalog No. p-7132603
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1214100MG 100 mg
M12141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1214100MG Supplier TCI America Supplier No. M1214100MG
Only null left

Chemical Identifiers

CAS 3189-13-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00022780
InChI Key QJRWYBIKLXNYLF-UHFFFAOYSA-N
Synonym 6-methoxyindole, 1h-indole, 6-methoxy, indole, 6-methoxy, 1h-indol-6-yl methyl ether, 6-methoxy indole, pubchem1701, acmc-209hph, opera_id_1903, 6-methyloxy-1h-indole
PubChem CID 76659
IUPAC Name 6-methoxy-1H-indole
SMILES COC1=CC=C2C=CNC2=C1

Specifications

Melting Point 93°C
Quantity 100 mg
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxyindole