missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxyindole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 93°C |
| Quantity | 1 g |
| Formula Weight | 147.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Methoxyindole |
Safety and Handling
| TSCA | No |