missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyindole, 98+%

Catalog No. AAL19074MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19074MD 250 mg
AAL1907403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19074MD Supplier Thermo Scientific Chemicals Supplier No. L19074MD
Only null left

CAS: 3189-13-7 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3189-13-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00022780
InChI Key QJRWYBIKLXNYLF-UHFFFAOYSA-N
Synonym 6-methoxyindole, 1h-indole, 6-methoxy, indole, 6-methoxy, 1h-indol-6-yl methyl ether, 6-methoxy indole, pubchem1701, acmc-209hph, opera_id_1903, 6-methyloxy-1h-indole
PubChem CID 76659
IUPAC Name 6-methoxy-1H-indole
SMILES COC1=CC=C2C=CNC2=C1

Specifications

Melting Point 90°C to 94°C
Boiling Point 105°C (0.2 mmHg)
Quantity 250 mg
Beilstein 116483
Sensitivity Light sensitive
Formula Weight 147.18
Percent Purity ≥98%
Chemical Name or Material 6-Methoxyindole
EINECSNumber 221-689-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only