missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyflavone 99.0+%, TCI America™

Catalog No. M13461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13461G 1 g
M13465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13461G Supplier TCI America Supplier No. M13461G
Only null left

Chemical Identifiers

CAS 26964-24-9
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.269
MDL Number MFCD00017322
InChI Key XZQLSABETMKIGG-UHFFFAOYSA-N
PubChem CID 147157
IUPAC Name 6-methoxy-2-phenylchromen-4-one
SMILES COC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 1 g
Synonym 6-methoxyflavone, 6-methoxy-2-phenyl-4h-chromen-4-one, 6-methoxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 6-methoxy-2-phenyl, 6-methoxy-2-phenyl-4h-1-benzopyran-4-one, 6-methoxy-2-phenyl-chromen-4-one, flavone, 6-methoxy-7ci,8ci, 4h-1-benzopyran-4-one,6-methoxy-2-phenyl, flavone, 6-methoxy
Formula Weight 252.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxyflavone
RTECSNumber DJ3100438
TSCA No