missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxybenzothiazole-2-carbonitrile, 99%

Catalog No. AAH5108206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5108206 5 g
AAH5108203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5108206 Supplier Thermo Scientific Chemicals Supplier No. H5108206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 943-03-3 | C9H6N2OS | 190.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 943-03-3
Molecular Formula C9H6N2OS
Molecular Weight (g/mol) 190.22
MDL Number MFCD00010537
InChI Key DEWDWBYQOFXKIH-UHFFFAOYSA-N
Synonym 2-cyano-6-methoxybenzothiazole, 6-methoxybenzo d thiazole-2-carbonitrile, 6-methoxy-2-benzothiazolecarbonitrile, 6-methoxybenzothiazole-2-carbonitrile, 6-methoxy-benzothiazole-2-carbonitrile, 6-methoxy-2-cyano-benzothiazole, 2-benzothiazolecarbonitrile, 6-methoxy, pubchem9766, acmc-209rr8, ksc486m5p
PubChem CID 342109
IUPAC Name 6-methoxy-1,3-benzothiazole-2-carbonitrile
SMILES COC1=CC2=C(C=C1)N=C(S2)C#N

Specifications

Quantity 5 g
Formula Weight 190.22
Percent Purity 99%
Chemical Name or Material 6-Methoxybenzothiazole-2-carbonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only