missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-4-methylcoumarin 98.0+%, TCI America™

Catalog No. p-7132706
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
M13985G 5 g
M139825G 25 g
2 options

Catalog No. M13985G

Supplier: TCI America M13985G

Only null left

Chemical Identifiers

CAS 6295-35-8
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00635101
InChI Key KNGPIBWCWOQBEK-UHFFFAOYSA-N
PubChem CID 223821
IUPAC Name 6-methoxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=C(C=C2)OC

Specifications

Melting Point 164°C
Quantity 5 g
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-4-methylcoumarin