missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-pyridinecarboxaldehyde 97.0+%, TCI America™

Catalog No. p-7133181
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23571G 1 g
M23575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23571G Supplier TCI America Supplier No. M23571G

Chemical Identifiers

CAS 54221-96-4
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD05664030
InChI Key YDNWTNODZDSPNZ-UHFFFAOYSA-N
Synonym 6-methoxypicolinaldehyde, 6-methoxy-2-pyridinecarboxaldehyde, 2-methoxy-6-pyridinecarboxaldehyde, 2-formyl-6-methoxypyridine, 6-methoxy-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-methoxy, 6-methoxypyridine-2-carboxaldehyde, pubchem15345, acmc-1aulb, ksc490s1l
PubChem CID 12325392
IUPAC Name 6-methoxypyridine-2-carbaldehyde
SMILES COC1=CC=CC(=N1)C=O

Specifications

Color Yellow
Boiling Point 104°C
Quantity 1 g
Formula Weight 137.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Methoxy-2-pyridinecarboxaldehyde