missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxy-2-naphthaldehyde 97.0+%, TCI America™
Supplier: TCI America M109025G
Specifications
| 6-Methoxy-2-naphthaldehyde | |
| 82°C | |
| C12H10O2 | |
| 25 g | |
| VZBLASFLFFMMCM-UHFFFAOYSA-N | |
| 6-methoxynaphthalene-2-carbaldehyde | |
| 76991 | |
| ≥97.0% (GC) |
| 3453-33-6 | |
| 196°C | |
| MFCD00081152 | |
| 6-methoxy-2-naphthaldehyde, 6-methoxy-2-naphthalaldehyde, unii-9244l5l5zy, 6-methoxy-naphthalene-2-carbaldehyde, 2-naphthalenecarboxaldehyde, 6-methoxy, pubchem13704, acmc-1ah4r, 6-methoxy-2-napthaldehyde, dsstox_cid_31510, dsstox_rid_97395 | |
| COC1=CC=C2C=C(C=O)C=CC2=C1 | |
| 186.21 | |
| 186.21 | |
| Crystalline Powder |
Chemical Identifiers
| 3453-33-6 | |
| 186.21 | |
| VZBLASFLFFMMCM-UHFFFAOYSA-N | |
| 76991 | |
| COC1=CC=C2C=C(C=O)C=CC2=C1 |
| C12H10O2 | |
| MFCD00081152 | |
| 6-methoxy-2-naphthaldehyde, 6-methoxy-2-naphthalaldehyde, unii-9244l5l5zy, 6-methoxy-naphthalene-2-carbaldehyde, 2-naphthalenecarboxaldehyde, 6-methoxy, pubchem13704, acmc-1ah4r, 6-methoxy-2-napthaldehyde, dsstox_cid_31510, dsstox_rid_97395 | |
| 6-methoxynaphthalene-2-carbaldehyde |
Safety and Handling
TSCA : No