missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-methylquinoline 98.0+%, TCI America™

Catalog No. p-7132715
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14145G 5 g
M141425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14145G Supplier TCI America Supplier No. M14145G
Only null left

Chemical Identifiers

CAS 1078-28-0
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD00006761
InChI Key NAGJQQFMJKMXJQ-UHFFFAOYSA-N
Synonym 6-methoxyquinaldine, 2-methyl-6-methoxyquinoline, quinoline, 6-methoxy-2-methyl, quinaldine, 6-methoxy, 6-methoxy-2-methyl-quinoline, quinoline,6-methoxy-2-methyl, pubchem5910, acmc-2098xp, 2-methy-6-methoxyquinolinel
PubChem CID 70648
IUPAC Name 6-methoxy-2-methylquinoline
SMILES CC1=NC2=C(C=C1)C=C(C=C2)OC

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 5 g
Formula Weight 173.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-2-methylquinoline