missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-methylquinoline, 97%

Catalog No. p-7026276
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2219903 1 g
AAB2219906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2219903 Supplier Thermo Scientific Chemicals Supplier No. B2219903

CAS: 1078-28-0 | C11H11NO | 173.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1078-28-0
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD00006761
InChI Key NAGJQQFMJKMXJQ-UHFFFAOYSA-N
Synonym 6-methoxyquinaldine, 2-methyl-6-methoxyquinoline, quinoline, 6-methoxy-2-methyl, quinaldine, 6-methoxy, 6-methoxy-2-methyl-quinoline, quinoline,6-methoxy-2-methyl, pubchem5910, acmc-2098xp, 2-methy-6-methoxyquinolinel
PubChem CID 70648
IUPAC Name 6-methoxy-2-methylquinoline
SMILES CC1=NC2=C(C=C1)C=C(C=C2)OC

Specifications

Melting Point 62°C to 66°C
Density 0.973
Flash Point 47°C (117°F)
Quantity 1 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material 6-Methoxy-2-methylquinoline
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 214-080-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only