missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2(3H)-benzoxazolone, 98+%

CAS: 532-91-2 | C8H7NO3 | 165.148 g/mol

Supplier:  Thermo Scientific Chemicals L0534903

 View more versions of this product

Catalog No. AAL0534903


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Methoxy-2(3H)-benzoxazolone
152°C to 156°C
MFCD00051726
141041
MKMCJLMBVKHUMS-UHFFFAOYSA-N
6-methoxy-3H-1,3-benzoxazol-2-one
10772
≥98%
532-91-2
C8H7NO3
1 g
coixol, 6-methoxy-2-benzoxazolinone, 6-methoxybenzo d oxazol-2 3h-one, 6-methoxybenzoxazolinone, mboa, 6-mboa, 6-methoxy-2 3h-benzoxazolone, 2-benzoxazolinone, 6-methoxy, 2 3h-benzoxazolone, 6-methoxy, 6-methoxy-1,3-benzoxazol-2 3h-one
COC1=CC2=C(C=C1)NC(=O)O2
165.148
165.15

Chemical Identifiers

532-91-2
165.148
MKMCJLMBVKHUMS-UHFFFAOYSA-N
10772
COC1=CC2=C(C=C1)NC(=O)O2
C8H7NO3
MFCD00051726
coixol, 6-methoxy-2-benzoxazolinone, 6-methoxybenzo d oxazol-2 3h-one, 6-methoxybenzoxazolinone, mboa, 6-mboa, 6-methoxy-2 3h-benzoxazolone, 2-benzoxazolinone, 6-methoxy, 2 3h-benzoxazolone, 6-methoxy, 6-methoxy-1,3-benzoxazol-2 3h-one
6-methoxy-3H-1,3-benzoxazol-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DM5275200

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only