missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2(3H)-benzoxazolone, 98+%

Catalog No. AAL05349MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL05349MD 250 mg
AAL0534903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL05349MD Supplier Thermo Scientific Chemicals Supplier No. L05349MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 532-91-2 | C8H7NO3 | 165.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 532-91-2
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00051726
InChI Key MKMCJLMBVKHUMS-UHFFFAOYSA-N
Synonym coixol, 6-methoxy-2-benzoxazolinone, 6-methoxybenzo d oxazol-2 3h-one, 6-methoxybenzoxazolinone, mboa, 6-mboa, 6-methoxy-2 3h-benzoxazolone, 2-benzoxazolinone, 6-methoxy, 2 3h-benzoxazolone, 6-methoxy, 6-methoxy-1,3-benzoxazol-2 3h-one
PubChem CID 10772
IUPAC Name 6-methoxy-3H-1,3-benzoxazol-2-one
SMILES COC1=CC2=C(C=C1)NC(=O)O2

Specifications

Melting Point 152°C to 156°C
Quantity 250 mg
Beilstein 141041
Formula Weight 165.15
Percent Purity ≥98%
Chemical Name or Material 6-Methoxy-2(3H)-benzoxazolone
RTECSNumber DM5275200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only