missing translation for 'onlineSavingsMsg'
Learn More

6'-Methoxy-2,3'-bipyridyl 97.0+%, TCI America™

Catalog No. p-7133262
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
M2509200MG 200 mg
M25091G 1 g
2 options

Catalog No. M2509200MG

Supplier: TCI America M2509200MG

Only null left

Chemical Identifiers

CAS 381725-49-1
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD11707142
InChI Key HRVJUCXZUHFRNP-UHFFFAOYSA-N
Synonym 6′C-Methoxy-2,3′C-bipyridine
PubChem CID 10899365
IUPAC Name 2-methoxy-5-pyridin-2-ylpyridine
SMILES COC1=NC=C(C=C1)C2=CC=CC=N2

Specifications

Color Yellow
Boiling Point 104°C
Quantity 200 mg
Formula Weight 186.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6′-Methoxy-2,3′-bipyridyl