missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Isopropylchromone-3-carbonitrile 98.0+%, TCI America™
$201.32 - $684.11
Chemical Identifiers
| CAS | 50743-32-3 |
|---|---|
| Molecular Formula | C13H11NO2 |
| Molecular Weight (g/mol) | 213.24 |
| MDL Number | MFCD00191961 |
| InChI Key | IMVAJLIIWCJMJP-UHFFFAOYSA-N |
| Synonym | 6-Isopropyl-4-oxo-4H-1-benzopyran-3-carbonitrile, 3-Cyano-6-isopropylchromone |
| PubChem CID | 736017 |
| IUPAC Name | 4-oxo-6-(propan-2-yl)-4H-chromene-3-carbonitrile |
| SMILES | CC(C)C1=CC=C2OC=C(C#N)C(=O)C2=C1 |
Chemical Identifiers
| 50743-32-3 | |
| 213.24 | |
| IMVAJLIIWCJMJP-UHFFFAOYSA-N | |
| 736017 | |
| CC(C)C1=CC=C2OC=C(C#N)C(=O)C2=C1 |
| C13H11NO2 | |
| MFCD00191961 | |
| 6-Isopropyl-4-oxo-4H-1-benzopyran-3-carbonitrile, 3-Cyano-6-isopropylchromone | |
| 4-oxo-6-(propan-2-yl)-4H-chromene-3-carbonitrile |
Specifications
| 50743-32-3 | |
| Yellow | |
| MFCD00191961 | |
| 6-Isopropyl-4-oxo-4H-1-benzopyran-3-carbonitrile, 3-Cyano-6-isopropylchromone | |
| CC(C)C1=CC=C2OC=C(C#N)C(=O)C2=C1 | |
| 213.24 | |
| 213.24 | |
| Crystalline Powder |
| 119°C | |
| C13H11NO2 | |
| 1 g | |
| IMVAJLIIWCJMJP-UHFFFAOYSA-N | |
| 4-oxo-6-(propan-2-yl)-4H-chromene-3-carbonitrile | |
| 736017 | |
| ≥98.0% (GC) | |
| 6-Isopropylchromone-3-carbonitrile |
Safety and Handling
TSCA : No