missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Isopropylchromone-3-carbonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 119°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 213.24 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Isopropylchromone-3-carbonitrile |
Safety and Handling
| TSCA | No |