missing translation for 'onlineSavingsMsg'
Learn More

6-Isopropylchromone-3-carbonitrile 98.0+%, TCI America™

Catalog No. C26611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26611G 1 g
C26615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26611G Supplier TCI America Supplier No. C26611G
Only null left

Chemical Identifiers

CAS 50743-32-3
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.24
MDL Number MFCD00191961
InChI Key IMVAJLIIWCJMJP-UHFFFAOYSA-N
Synonym 6-Isopropyl-4-oxo-4H-1-benzopyran-3-carbonitrile, 3-Cyano-6-isopropylchromone
PubChem CID 736017
IUPAC Name 4-oxo-6-(propan-2-yl)-4H-chromene-3-carbonitrile
SMILES CC(C)C1=CC=C2OC=C(C#N)C(=O)C2=C1

Specifications

Melting Point 119°C
Color Yellow
Quantity 1 g
Formula Weight 213.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Isopropylchromone-3-carbonitrile