missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo-1H-indazole, 97%, Thermo Scientific™

Catalog No. AC432420010
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432420010 1g Glass bottle
AC432420050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432420010 Supplier Thermo Scientific Chemicals Supplier No. 432420010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 261953-36-0
Molecular Formula C7H5IN2
Molecular Weight (g/mol) 244.03
MDL Number MFCD04114695
InChI Key RSGAXJZKQDNFEP-UHFFFAOYSA-N
Synonym 6-iodoindazole, 1h-indazole, 6-iodo, 6-iodo 1h indazole, 6-iodo-indazole, pubchem18081, indazole, 6-iodo
PubChem CID 12991241
IUPAC Name 6-iodo-1H-indazole
SMILES C1=CC2=C(C=C1I)NN=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1g
Formula Weight 244.03
Percent Purity 97%
Chemical Name or Material 6-Iodo-1H-indazole
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only