missing translation for 'onlineSavingsMsg'
Learn More

6-Indoleboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. I08031G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08031G Supplier TCI America Supplier No. I08031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 147621-18-9
Molecular Formula C8H8BNO2
Molecular Weight (g/mol) 160.97
MDL Number MFCD03095176
InChI Key ZVMHOIWRCCZGPZ-UHFFFAOYSA-N
Synonym indole-6-boronic acid, 6-indolylboronic acid, 1h-indol-6-yl boronic acid, 6-indoleboronic acid, indole-6-boronicacid, 1h-indol-6-yl-6-boronic acid, 6-borono-1h-indole, 6-indole boronic acid, 6-boronoindole, 1h-indole-6-boronic acid
PubChem CID 2763205
IUPAC Name (1H-indol-6-yl)boronic acid
SMILES OB(O)C1=CC=C2C=CNC2=C1

Specifications

Melting Point 200°C
Color Yellow
Quantity 1 g
Formula Weight 160.97
Physical Form Crystalline Powder
Chemical Name or Material 6-Indoleboronic Acid (contains varying amounts of Anhydride)
TSCA No
Recommended Storage Refrigerator