missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxypyridine-2-carboxylic acid, 95%

Catalog No. AC434820050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434820050 5 g Glass bottle
AC434820250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434820050 Supplier Thermo Scientific Chemicals Supplier No. 434820050

CAS: 19621-92-2 | C6H5NO3 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19621-92-2
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00192220
InChI Key VRCWSYYXUCKEED-UHFFFAOYSA-N
Synonym 6-hydroxypicolinic acid, 6-hydroxy-2-pyridinecarboxylic acid, 6-hydroxypyridine-2-carboxylic acid, 6-oxo-1,6-dihydropyridine-2-carboxylic acid, 6-carboxy-2-hydroxy-pyridine, 6-hydroxy-2-pyridine carboxylic acid, 6-hydroxy-2-pyridinecarboxylicacid, 2-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylicacid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylic acid,1,6-dihydro-6-oxo
PubChem CID 242721
IUPAC Name 6-oxo-1H-pyridine-2-carboxylic acid
SMILES C1=CC(=O)NC(=C1)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 273°C to 276°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 139.11
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 6-Hydroxypyridine-2-carboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

RUO – Research Use Only