missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyindole, 98%

Catalog No. p-7037255
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55951MD 250 mg
AAH5595103 1 g
AAH5595106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55951MD Supplier Thermo Scientific Chemicals Supplier No. H55951MD
Only null left

CAS: 2380-86-1 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2380-86-1
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00152101
InChI Key XAWPKHNOFIWWNZ-UHFFFAOYSA-N
Synonym 6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol
PubChem CID 524508
IUPAC Name 1H-indol-6-ol
SMILES C1=CC(=CC2=C1C=CN2)O

Specifications

Melting Point 125°C to 128°C
Quantity 250 mg
UN Number UN3077
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 6-Hydroxyindole
Hazard Category H302-H317-H318
Hazard Statement GHS H Statement
H318-H302-H317
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 417-020-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only