Learn More
6-Hydroxyindole, 97%
CAS: 2380-86-1 | C8H7NO | 133.15 g/mol
$424.64 - $424.64
Chemical Identifiers
| CAS | 2380-86-1 |
|---|---|
| Molecular Formula | C8H7NO |
| Molecular Weight (g/mol) | 133.15 |
| InChI Key | XAWPKHNOFIWWNZ-UHFFFAOYSA-N |
| Synonym | 6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol |
| PubChem CID | 524508 |
| IUPAC Name | 1H-indol-6-ol |
| SMILES | C1=CC(=CC2=C1C=CN2)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC379010010
|
Thermo Scientific Chemicals
379010010 |
1 g | Glass bottle |
Each for $424.64
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2380-86-1 | |
| 133.15 | |
| 6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol | |
| 1H-indol-6-ol |
| C8H7NO | |
| XAWPKHNOFIWWNZ-UHFFFAOYSA-N | |
| 524508 | |
| C1=CC(=CC2=C1C=CN2)O |
Specifications
| 2380-86-1 | |
| White | |
| 97.5% min. (HPLC) | |
| C8H7NO | |
| 6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol | |
| C1=CC(=CC2=C1C=CN2)O | |
| 133.15 | |
| 133.15 | |
| Shiny Crystalline Powder |
| 124.0°C to 130.0°C | |
| Authentic | |
| Glass bottle | |
| 1 g | |
| XAWPKHNOFIWWNZ-UHFFFAOYSA-N | |
| 1H-indol-6-ol | |
| 524508 | |
| 97% | |
| 6-Hydroxyindole |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protecti
GHS Signal Word: Danger
Recommended Storage : Product may darken during storage
RUO – Research Use Only