missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyhexanoic acid, 95%, may cont. variable amounts of dimer

Catalog No. AAB2485714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2485714 25 g
AAB2485703 1 g
AAB2485706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2485714 Supplier Thermo Scientific Chemicals Supplier No. B2485714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1191-25-9 | C6H12O3 | 132.159 g/mol

6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1191-25-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00046560
InChI Key IWHLYPDWHHPVAA-UHFFFAOYSA-N
Synonym 6-hydroxycaproic acid, 6-hydroxyhexanoate, hexanoic acid, 6-hydroxy, 6-hydroxy caproic acid, 6-hydroxy-hexanoic acid, epsilon-hydroxycaproic acid, 5-carboxypentanol, unii-3y3ox37nm8, 6-hydroxycaproicacid,pract.,containslactone, 6-hydroxy-caproic acid
PubChem CID 14490
ChEBI CHEBI:17869
IUPAC Name 6-hydroxyhexanoic acid
SMILES C(CCC(=O)O)CCO

Specifications

Melting Point 38°C to 40°C
Density 1.08
Flash Point >110°C (230°F)
Refractive Index 1.456
Linear Formula HO(CH2)5CO2H
Quantity 25 g
Formula Weight 132.16
Percent Purity 95%
Assay may cont. variable amounts of dimer
Chemical Name or Material 6-Hydroxyhexanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only