missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyflavone 98.0+%, TCI America™

$89.48 - $216.24

Chemical Identifiers

CAS 6665-83-4
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.242
MDL Number MFCD00017329
InChI Key GPZYYYGYCRFPBU-UHFFFAOYSA-N
Synonym 6-hydroxyflavone, 6-monohydroxyflavone, 6-hydroxy-2-phenyl-4-benzopyrone, 6-hydroxy-2-phenyl-chromen-4-one, 6-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 6-hydroxy-2-phenyl, 6-hydroxy-flavone, 6-hydroxy flavone, 6-hydroxyflavone, 10
PubChem CID 72279
ChEBI CHEBI:34472
IUPAC Name 6-hydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC(=C3)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08511G
View Documents
TCI America
H08511G
1 g
Each for $89.48
Only null left
 
H08515G
View Documents
TCI America
H08515G
5 g
Each for $216.24
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6665-83-4
238.242
GPZYYYGYCRFPBU-UHFFFAOYSA-N
72279
6-hydroxy-2-phenylchromen-4-one
C15H10O3
MFCD00017329
6-hydroxyflavone, 6-monohydroxyflavone, 6-hydroxy-2-phenyl-4-benzopyrone, 6-hydroxy-2-phenyl-chromen-4-one, 6-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 6-hydroxy-2-phenyl, 6-hydroxy-flavone, 6-hydroxy flavone, 6-hydroxyflavone, 10
CHEBI:34472
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC(=C3)O

Specifications

6665-83-4
MFCD00017329
6-hydroxyflavone, 6-monohydroxyflavone, 6-hydroxy-2-phenyl-4-benzopyrone, 6-hydroxy-2-phenyl-chromen-4-one, 6-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 6-hydroxy-2-phenyl, 6-hydroxy-flavone, 6-hydroxy flavone, 6-hydroxyflavone, 10
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC(=C3)O
238.242
CHEBI:34472
≥98.0% (T,HPLC)
6-Hydroxyflavone
C15H10O3
1 g
GPZYYYGYCRFPBU-UHFFFAOYSA-N
6-hydroxy-2-phenylchromen-4-one
72279
238.24
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'