missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyflavone 98.0+%, TCI America™

Catalog No. H08511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08511G 1 g
H08515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08511G Supplier TCI America Supplier No. H08511G

Chemical Identifiers

CAS 6665-83-4
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.242
MDL Number MFCD00017329
InChI Key GPZYYYGYCRFPBU-UHFFFAOYSA-N
Synonym 6-hydroxyflavone, 6-monohydroxyflavone, 6-hydroxy-2-phenyl-4-benzopyrone, 6-hydroxy-2-phenyl-chromen-4-one, 6-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 6-hydroxy-2-phenyl, 6-hydroxy-flavone, 6-hydroxy flavone, 6-hydroxyflavone, 10
PubChem CID 72279
ChEBI CHEBI:34472
IUPAC Name 6-hydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC(=C3)O

Specifications

Quantity 1 g
Formula Weight 238.24
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxyflavone