missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxyflavanone 98.0+%, TCI America™
$93.21 - $303.84
Chemical Identifiers
| CAS | 4250-77-5 |
|---|---|
| Molecular Formula | C15H12O3 |
| Molecular Weight (g/mol) | 240.258 |
| MDL Number | MFCD00017485 |
| InChI Key | XYHWPQUEOOBIOW-UHFFFAOYSA-N |
| Synonym | 6-hydroxyflavanone, 6-hydroxy-2-phenylchroman-4-one, 6-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-hydroxy-flavanone, 4h-1-benzopyran-4-one, 2,3-dihydro-6-hydroxy-2-phenyl, 6-hydroxy-2-phenyl-chroman-4-one, flavanone, 6-hydroxy, +/--6-hydroxyflavanone, acmc-209jpu |
| PubChem CID | 2734580 |
| ChEBI | CHEBI:34471 |
| IUPAC Name | 6-hydroxy-2-phenyl-2,3-dihydrochromen-4-one |
| SMILES | C1C(OC2=C(C1=O)C=C(C=C2)O)C3=CC=CC=C3 |
Chemical Identifiers
| 4250-77-5 | |
| 240.258 | |
| XYHWPQUEOOBIOW-UHFFFAOYSA-N | |
| 2734580 | |
| 6-hydroxy-2-phenyl-2,3-dihydrochromen-4-one |
| C15H12O3 | |
| MFCD00017485 | |
| 6-hydroxyflavanone, 6-hydroxy-2-phenylchroman-4-one, 6-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-hydroxy-flavanone, 4h-1-benzopyran-4-one, 2,3-dihydro-6-hydroxy-2-phenyl, 6-hydroxy-2-phenyl-chroman-4-one, flavanone, 6-hydroxy, +/--6-hydroxyflavanone, acmc-209jpu | |
| CHEBI:34471 | |
| C1C(OC2=C(C1=O)C=C(C=C2)O)C3=CC=CC=C3 |
Specifications
| 4250-77-5 | |
| White-Yellow | |
| MFCD00017485 | |
| 6-hydroxyflavanone, 6-hydroxy-2-phenylchroman-4-one, 6-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-hydroxy-flavanone, 4h-1-benzopyran-4-one, 2,3-dihydro-6-hydroxy-2-phenyl, 6-hydroxy-2-phenyl-chroman-4-one, flavanone, 6-hydroxy, +/--6-hydroxyflavanone, acmc-209jpu | |
| C1C(OC2=C(C1=O)C=C(C=C2)O)C3=CC=CC=C3 | |
| 240.258 | |
| CHEBI:34471 | |
| ≥98.0% (GC,T) | |
| 6-Hydroxyflavanone |
| 219°C | |
| C15H12O3 | |
| 1 g | |
| XYHWPQUEOOBIOW-UHFFFAOYSA-N | |
| 6-hydroxy-2-phenyl-2,3-dihydrochromen-4-one | |
| 2734580 | |
| 240.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No