missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyflavanone 98.0+%, TCI America™

Catalog No. H10271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10271G 1 g
H10275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10271G Supplier TCI America Supplier No. H10271G

Chemical Identifiers

CAS 4250-77-5
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.258
MDL Number MFCD00017485
InChI Key XYHWPQUEOOBIOW-UHFFFAOYSA-N
Synonym 6-hydroxyflavanone, 6-hydroxy-2-phenylchroman-4-one, 6-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-hydroxy-flavanone, 4h-1-benzopyran-4-one, 2,3-dihydro-6-hydroxy-2-phenyl, 6-hydroxy-2-phenyl-chroman-4-one, flavanone, 6-hydroxy, +/--6-hydroxyflavanone, acmc-209jpu
PubChem CID 2734580
ChEBI CHEBI:34471
IUPAC Name 6-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
SMILES C1C(OC2=C(C1=O)C=C(C=C2)O)C3=CC=CC=C3

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 1 g
Formula Weight 240.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxyflavanone