missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-5-nitronicotinic Acid 97.0+%, TCI America™

Catalog No. H14105G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14105G 5 g
H14101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14105G Supplier TCI America Supplier No. H14105G

Chemical Identifiers

CAS 6635-31-0
Molecular Formula C6H4N2O5
Molecular Weight (g/mol) 184.107
MDL Number MFCD02180840
InChI Key VJMXJGVEVASJOD-UHFFFAOYSA-N
Synonym 6-hydroxy-5-nitronicotinic acid, 6-hydroxy-5-nitropyridine-3-carboxylic acid, 6-hydroxy-5-nitronicotinicacid, 5-nitro-6-oxo-1h-pyridine-3-carboxylic acid, 6-hydroxy-5-nitro-nicotinic acid, 5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylic acid, 6-hydroxy-5-nitro nicotinic acid, 3-pyridinecarboxylicacid, 1,6-dihydro-5-nitro-6-oxo, 3-pyridinecarboxylic acid, 1,6-dihydro-5-nitro-6-oxo, pubchem14325
PubChem CID 243019
SMILES C1=C(C(=O)NC=C1C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 291°C
Color Yellow
Quantity 5 g
Formula Weight 184.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-5-nitronicotinic Acid