missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxy-2-methyl-3-nitropyridine 98.0+%, TCI America™
$54.06 - $160.30
Chemical Identifiers
| CAS | 28489-45-4 |
|---|---|
| Molecular Formula | C6H6N2O3 |
| Molecular Weight (g/mol) | 154.125 |
| InChI Key | AJDDHNXZBGZBJN-UHFFFAOYSA-N |
| Synonym | 6-methyl-5-nitropyridin-2-ol, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 2-hydroxy-5-nitro-6-picoline, 6-hydroxy-2-methyl-3-nitropyridine, 6-methyl-5-nitropyridin-2 1h-one, 2 1h-pyridinone, 6-methyl-5-nitro, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, pubchem10387 |
| PubChem CID | 543053 |
| IUPAC Name | 6-methyl-5-nitro-1H-pyridin-2-one |
| SMILES | CC1=C(C=CC(=O)N1)[N+](=O)[O-] |
Chemical Identifiers
| 28489-45-4 | |
| 154.125 | |
| 6-methyl-5-nitropyridin-2-ol, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 2-hydroxy-5-nitro-6-picoline, 6-hydroxy-2-methyl-3-nitropyridine, 6-methyl-5-nitropyridin-2 1h-one, 2 1h-pyridinone, 6-methyl-5-nitro, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, pubchem10387 | |
| 6-methyl-5-nitro-1H-pyridin-2-one |
| C6H6N2O3 | |
| AJDDHNXZBGZBJN-UHFFFAOYSA-N | |
| 543053 | |
| CC1=C(C=CC(=O)N1)[N+](=O)[O-] |
Specifications
| 28489-45-4 | |
| C6H6N2O3 | |
| 6-methyl-5-nitropyridin-2-ol, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 2-hydroxy-5-nitro-6-picoline, 6-hydroxy-2-methyl-3-nitropyridine, 6-methyl-5-nitropyridin-2 1h-one, 2 1h-pyridinone, 6-methyl-5-nitro, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, pubchem10387 | |
| CC1=C(C=CC(=O)N1)[N+](=O)[O-] | |
| 154.125 | |
| 154.13 | |
| Crystalline Powder |
| 236°C | |
| 1 g | |
| AJDDHNXZBGZBJN-UHFFFAOYSA-N | |
| 6-methyl-5-nitro-1H-pyridin-2-one | |
| 543053 | |
| ≥98.0% (GC,T) | |
| 6-Hydroxy-2-methyl-3-nitropyridine |
Safety and Handling
TSCA : No