missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-methyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. H11605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11605G 5 g
H11601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11605G Supplier TCI America Supplier No. H11605G

Chemical Identifiers

CAS 28489-45-4
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
InChI Key AJDDHNXZBGZBJN-UHFFFAOYSA-N
Synonym 6-methyl-5-nitropyridin-2-ol, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 2-hydroxy-5-nitro-6-picoline, 6-hydroxy-2-methyl-3-nitropyridine, 6-methyl-5-nitropyridin-2 1h-one, 2 1h-pyridinone, 6-methyl-5-nitro, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, pubchem10387
PubChem CID 543053
IUPAC Name 6-methyl-5-nitro-1H-pyridin-2-one
SMILES CC1=C(C=CC(=O)N1)[N+](=O)[O-]

Specifications

Melting Point 236°C
Quantity 5 g
Formula Weight 154.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-2-methyl-3-nitropyridine