missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich (+/-)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

Catalog No. 2388135G
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2388135G 5 g
2388131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2388135G Supplier Sigma Aldrich Supplier No. 2388135G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+/-)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

Specifications

CAS 53188-07-1
Melting Point 187°C to 189°C (lit.)
Molecular Formula C14H18O4
Linear Formula C14H18O4
MDL Number MFCD00006846
Quantity 5 g
Synonym Trolox
Molecular Weight (g/mol) 250.29
Percent Purity 97%