missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich (+/-)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid
Description
(+/-)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid
Specifications
Specifications
| CAS | 53188-07-1 |
| Melting Point | 187°C to 189°C (lit.) |
| Molecular Formula | C14H18O4 |
| Linear Formula | C14H18O4 |
| MDL Number | MFCD00006846 |
| Quantity | 1 g |
| Synonym | Trolox |
| Molecular Weight (g/mol) | 250.29 |
| Percent Purity | 97% |