missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. H07261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07261G 1 g
H07265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07261G Supplier TCI America Supplier No. H07261G

Chemical Identifiers

CAS 53188-07-1
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.29
MDL Number MFCD00006846
InChI Key GLEVLJDDWXEYCO-UHFFFAOYNA-N
Synonym trolox, 6-hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, trolox c, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-2h-1-benzopyran-2-carboxylic acid, 6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl, +/--6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-, +-, trolox™, trolox tm
PubChem CID 40634
ChEBI CHEBI:82625
IUPAC Name 6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid
SMILES CC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 1 g
Formula Weight 250.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic Acid
RTECSNumber DJ2273000
TSCA Yes