missing translation for 'onlineSavingsMsg'
Learn More

6-Heptynoic acid, 95%

CAS: 30964-00-2 | C7H10O2 | 126.16 g/mol

$85.14 - $1,209.96

Chemical Identifiers

CAS 30964-00-2
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00239430
InChI Key OFCPMJGTZUVUSM-UHFFFAOYSA-N
Synonym 6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa
PubChem CID 4377950
IUPAC Name hept-6-ynoic acid
SMILES OC(=O)CCCCC#C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH5351903
View Documents Promotion Details
N/A 1 g
Each for $85.14
Only null left
 
AAH5351906
View Documents Promotion Details
N/A 5 g
Each for $269.85
Only null left
 
AAH5351914
View Documents Promotion Details
N/A 25 g
Each for $1,209.96
Only null left
 
Description

Description

6-Heptynoic acid is used as a precursor for the the preparation of boradiazaindacenes (BODIPY dyes) by reacting with oxalylchloirde. It is used to prepare 4,4-difluoro-8-(hept-6-yne)-3,5-di(2-(4-methoxyphenyl))-4-bora-3a,4a-diaza-sindacene by reacting with 2-(4-methoxyphenyl)-1H-pyrrole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Heptynoic acid is used as a precursor for the the preparation of boradiazaindacenes (BODIPY dyes) by reacting with oxalylchloirde. It is used to prepare 4,4-difluoro-8-(hept-6-yne)-3,5-di(2-(4-methoxyphenyl))-4-bora-3a,4a-diaza-sindacene by reacting with 2-(4-methoxyphenyl)-1H-pyrrole.

Solubility
Miscible with dimethylformamide.

Notes
Incompatible with oxidizing agents and bases
Specifications

Chemical Identifiers

30964-00-2
126.16
OFCPMJGTZUVUSM-UHFFFAOYSA-N
4377950
OC(=O)CCCCC#C
C7H10O2
MFCD00239430
6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa
hept-6-ynoic acid

Specifications

30964-00-2
93°C to 94°C (1 mmHg)
C7H10O2
HC≡C(CH2)4CO2H
UN3265
6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa
OFCPMJGTZUVUSM-UHFFFAOYSA-N
hept-6-ynoic acid
4377950
95%
0.997
113°C (235°F)
1.451
MFCD00239430
1747024
Miscible with dimethylformamide.
OC(=O)CCCCC#C
126.16
126.16
6-Heptynoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only