Learn More
6-Heptyn-1-ol, 95%
CAS: 63478-76-2 | C7H12O | 112.17 g/mol
Supplier: Thermo Scientific Chemicals H6150906
Description
6-Heptyn-1-ol is used as a reagent in in the preparation of many cyclic compounds. It is also used as a Intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Heptyn-1-ol is used as a reagent in in the preparation of many cyclic compounds. It is also used as a Intermediate in organic synthesis.
Solubility
Soluble in chloroform, dichloromethane and methanol. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
| 6-Heptyn-1-ol | |
| C7H12O | |
| MFCD00049198 | |
| 6-heptyn-1-ol, 7-hydroxyhept-1-yne, pubchem13080, ho ch2 5c=ch, ksc352m8p | |
| BVRCLEXKQNWTDK-UHFFFAOYSA-N | |
| hept-6-yn-1-ol | |
| 11007849 | |
| 95% |
| 63478-76-2 | |
| HC≡C(CH2)5OH | |
| 5 g | |
| Soluble in chloroform,dichloromethane and methanol. Slightly soluble in water. | |
| OCCCCCC#C | |
| 112.17 | |
| 112.17 |
Chemical Identifiers
| 63478-76-2 | |
| 112.17 | |
| BVRCLEXKQNWTDK-UHFFFAOYSA-N | |
| 11007849 | |
| OCCCCCC#C |
| C7H12O | |
| MFCD00049198 | |
| 6-heptyn-1-ol, 7-hydroxyhept-1-yne, pubchem13080, ho ch2 5c=ch, ksc352m8p | |
| hept-6-yn-1-ol |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only