missing translation for 'onlineSavingsMsg'
Learn More

6-Heptenoic acid, 96%

Catalog No. p-7046108
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1314403 1 g
AAL1314406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1314403 Supplier Thermo Scientific Chemicals Supplier No. L1314403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1119-60-4 | C7H12O2 | 128.171 g/mol

6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 1119-60-4
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00014382
InChI Key RWNJOXUVHRXHSD-UHFFFAOYSA-N
Synonym 6-heptenoic acid, epsilon-heptenoic acid, rarechem al bo 1361, 6-heptenoic acids, hept-6-enoic acids, acmc-20alyp, epsilon-heptenoic acids, 6-heptenoic acid 1g
PubChem CID 70705
ChEBI CHEBI:38363
IUPAC Name hept-6-enoic acid
SMILES C=CCCCCC(=O)O

Specifications

Melting Point ∼-6°C
Density 0.946
Boiling Point 222°C to 224°C
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.439
Quantity 1 g
UN Number UN3265
Beilstein 1747265
Solubility Information Miscible with water.
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material 6-Heptenoic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 214-283-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only