missing translation for 'onlineSavingsMsg'
Learn More

6-Heptenenitrile, 96%

CAS: 5048-25-9 | C7H11N | 109.17 g/mol

$333.64 - $333.64

Chemical Identifiers

CAS 5048-25-9
Molecular Formula C7H11N
Molecular Weight (g/mol) 109.17
MDL Number MFCD00151804
InChI Key ZLWQKLRMIQIHKQ-UHFFFAOYSA-N
Synonym 6-heptenenitrile, 6-cyano-1-hexene, acmc-20alyq
PubChem CID 4140856
IUPAC Name hept-6-enenitrile
SMILES C=CCCCCC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1313706
View Documents
Thermo Scientific Chemicals
L1313706
5 g
Each for $333.64
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5048-25-9
109.17
ZLWQKLRMIQIHKQ-UHFFFAOYSA-N
4140856
C=CCCCCC#N
C7H11N
MFCD00151804
6-heptenenitrile, 6-cyano-1-hexene, acmc-20alyq
hept-6-enenitrile

Specifications

5048-25-9
>100°C (212°F)
1.433
MFCD00151804
UN3276
6-heptenenitrile, 6-cyano-1-hexene, acmc-20alyq
C=CCCCCC#N
109.17
109.17
6-Heptenenitrile
75°C to 78°C (13 mmHg)
C7H11N
CH2=CH(CH2)4CN
5 g
1740406
ZLWQKLRMIQIHKQ-UHFFFAOYSA-N
hept-6-enenitrile
4140856
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only