missing translation for 'onlineSavingsMsg'
Learn More

6-Heptenenitrile, 96%

Catalog No. AAL1313706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1313706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1313706 Supplier Thermo Scientific Chemicals Supplier No. L1313706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5048-25-9 | C7H11N | 109.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5048-25-9
Molecular Formula C7H11N
Molecular Weight (g/mol) 109.17
MDL Number MFCD00151804
InChI Key ZLWQKLRMIQIHKQ-UHFFFAOYSA-N
Synonym 6-heptenenitrile, 6-cyano-1-hexene, acmc-20alyq
PubChem CID 4140856
IUPAC Name hept-6-enenitrile
SMILES C=CCCCCC#N

Specifications

Boiling Point 75°C to 78°C (13 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.433
Linear Formula CH2=CH(CH2)4CN
Quantity 5 g
UN Number UN3276
Beilstein 1740406
Formula Weight 109.17
Percent Purity 97%
Chemical Name or Material 6-Heptenenitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only