missing translation for 'onlineSavingsMsg'
Learn More

Code Corporation™ 6 ft. Straight RS232 Cable

Catalog No. 17579726
Change view
Click to view available options
For Use With (Equipment):
Code Readers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17579726 Code Readers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17579726 Supplier Code Corporation™ Supplier No. 176701

Use this 6 ft. Straight RS232 Affinity™ Cable CRA-C511 with Code™'s cabled barcode readers.

  • Reader termination: 10-pin RJ45
  • Host termination: 9-pin, female, D-Sub connector

Specifications

For Use With (Equipment) Code Readers
Type Cable
Alternate Part Number CRA-C502