missing translation for 'onlineSavingsMsg'
Learn More

Labconco 6 ft. Nexus Horizontal Clean Bench

Catalog No. 17195072
Change view
Click to view available options
Material:
Epoxy-coated Steel
Stainless Steel
Voltage:
115 V
230 V
Includes:
Base Stand
Clean Bench Only
UV Lamp
UV Lamp and Base Stand
Depth (English) Interior:
21 in.
26 in.
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Voltage Includes Depth (English) Interior
17195072 Epoxy-coated Steel 115 V Clean Bench Only 21 in.
17194987 Epoxy-coated Steel 230 V Clean Bench Only 21 in.
17195093 Stainless Steel 115 V Clean Bench Only 21 in.
17195036 Stainless Steel 230 V Clean Bench Only 21 in.
17195086 Epoxy-coated Steel 115 V UV Lamp 21 in.
17194976 Epoxy-coated Steel 230 V UV Lamp 21 in.
17194983 Stainless Steel 115 V UV Lamp 21 in.
17195079 Stainless Steel 230 V UV Lamp 21 in.
17195077 Epoxy-coated Steel 115 V Base Stand 21 in.
17194984 Epoxy-coated Steel 230 V Base Stand 21 in.
17194995 Stainless Steel 115 V Base Stand 21 in.
17195073 Stainless Steel 230 V Base Stand 21 in.
17194985 Epoxy-coated Steel 115 V UV Lamp and Base Stand 21 in.
17195022 Epoxy-coated Steel 230 V UV Lamp and Base Stand 21 in.
17195048 Stainless Steel 115 V UV Lamp and Base Stand 21 in.
17194955 Stainless Steel 230 V UV Lamp and Base Stand 21 in.
17194993 Epoxy-coated Steel 115 V Clean Bench Only 26 in.
17195016 Epoxy-coated Steel 230 V Clean Bench Only 26 in.
17195074 Stainless Steel 115 V Clean Bench Only 26 in.
17195007 Stainless Steel 230 V Clean Bench Only 26 in.
17195034 Epoxy-coated Steel 115 V UV Lamp 26 in.
17195003 Epoxy-coated Steel 230 V UV Lamp 26 in.
17194969 Stainless Steel 115 V UV Lamp 26 in.
17194968 Stainless Steel 230 V UV Lamp 26 in.
17194971 Epoxy-coated Steel 115 V Base Stand 26 in.
17195037 Epoxy-coated Steel 230 V Base Stand 26 in.
17194954 Stainless Steel 115 V Base Stand 26 in.
17195025 Stainless Steel 230 V Base Stand 26 in.
17195071 Epoxy-coated Steel 115 V UV Lamp and Base Stand 26 in.
17194986 Epoxy-coated Steel 230 V UV Lamp and Base Stand 26 in.
17195087 Stainless Steel 115 V UV Lamp and Base Stand 26 in.
17194970 Stainless Steel 230 V UV Lamp and Base Stand 26 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
Catalog No. 17195072 Supplier Labconco Supplier No. 5360000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.

  • 6 ft. Nexus Horizontal Clean Bench
  • ISO Class 4 laminar airflow is 10x cleaner than the industry standard ISO Class 5
  • Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications

Specifications

Product Type Horizontal Clean Bench
Coating Epoxy
Filter Type HEPA
Lighting Type LED
Material Epoxy-coated Steel
Color Optic White
Depth (English) Interior 21 in.
Depth (Metric) Interior 53.8 cm
Height (English) 50.1 in.
Height (English) Interior 30.2 in.
Height (Metric) 127.5 cm
Height (Metric) Interior 76.7 cm
Includes Clean Bench Only
IP Rating None
Plug Type NEMA 5-20P
Voltage 115 V
Width (English) 75.2 in.
Width (English) Interior 73.0 in
Width (Metric) 191.1 cm
Width (Metric) Interior 185.4 cm
Show More Show Less