missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco 6 ft. Nexus Horizontal Clean Bench
Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.
Supplier: Labconco 5361700
Description
- 6 ft. Nexus Horizontal Clean Bench
- ISO Class 4 laminar airflow is 10x cleaner than the industry standard ISO Class 5
- Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications
Chemical Identifiers
| Horizontal Clean Bench | |
| HEPA | |
| 50.1 in. | |
| 127.5 cm | |
| None | |
| 75.2 in. | |
| 191.1 cm |
| Epoxy | |
| Optic White | |
| 30.2 in. | |
| 76.7 cm | |
| NEMA 5-20P | |
| 73.0 in | |
| 185.4 cm |
Specifications
| Horizontal Clean Bench | |
| HEPA | |
| Epoxy-coated Steel | |
| 26 in. | |
| 50.1 in. | |
| 127.5 cm | |
| UV Lamp and Base Stand | |
| NEMA 5-20P | |
| 75.2 in. | |
| 191.1 cm |
| Epoxy | |
| UV | |
| Optic White | |
| 66.6 cm | |
| 30.2 in. | |
| 76.7 cm | |
| None | |
| 115 V | |
| 73.0 in | |
| 185.4 cm |