missing translation for 'onlineSavingsMsg'
Learn More

Labconco 6 ft. Nexus Horizontal Clean Bench

Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.

Supplier:  Labconco 5361500

 View more versions of this product

Catalog No. 17194971

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • 6 ft. Nexus Horizontal Clean Bench
  • ISO Class 4 laminar airflow is 10x cleaner than the industry standard ISO Class 5
  • Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications
Specifications

Chemical Identifiers

Horizontal Clean Bench
HEPA
50.1 in.
127.5 cm
None
75.2 in.
191.1 cm
Epoxy
Optic White
30.2 in.
76.7 cm
NEMA 5-20P
73.0 in
185.4 cm

Specifications

Horizontal Clean Bench
HEPA
Epoxy-coated Steel
26 in.
50.1 in.
127.5 cm
Base Stand
NEMA 5-20P
75.2 in.
191.1 cm
Epoxy
LED
Optic White
66.6 cm
30.2 in.
76.7 cm
None
115 V
73.0 in
185.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions