missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluorosalicylaldehyde 98.0+%, TCI America™
$234.87 - $829.75
Chemical Identifiers
| CAS | 38226-10-7 |
|---|---|
| Molecular Formula | C7H5FO2 |
| Molecular Weight (g/mol) | 140.113 |
| MDL Number | MFCD01090996 |
| InChI Key | FZIBGCDUHZBOLA-UHFFFAOYSA-N |
| Synonym | 6-fluorosalicylaldehyde, 3-fluoro-2-formylphenol, benzaldehyde, 2-fluoro-6-hydroxy, 6-fluoro-2-hydroxybenzaldehyde, pubchem8482, acmc-20a0l1, 6-fluorosalicyaldehyde, ksc497m7f, 2-hydroxy-6-fluorobenzaldehyde, 2-fluoro-6-hydroxy-benzaldehyde |
| PubChem CID | 2737327 |
| IUPAC Name | 2-fluoro-6-hydroxybenzaldehyde |
| SMILES | C1=CC(=C(C(=C1)F)C=O)O |
Chemical Identifiers
| 38226-10-7 | |
| 140.113 | |
| FZIBGCDUHZBOLA-UHFFFAOYSA-N | |
| 2737327 | |
| C1=CC(=C(C(=C1)F)C=O)O |
| C7H5FO2 | |
| MFCD01090996 | |
| 6-fluorosalicylaldehyde, 3-fluoro-2-formylphenol, benzaldehyde, 2-fluoro-6-hydroxy, 6-fluoro-2-hydroxybenzaldehyde, pubchem8482, acmc-20a0l1, 6-fluorosalicyaldehyde, ksc497m7f, 2-hydroxy-6-fluorobenzaldehyde, 2-fluoro-6-hydroxy-benzaldehyde | |
| 2-fluoro-6-hydroxybenzaldehyde |
Specifications
| 38226-10-7 | |
| C7H5FO2 | |
| 1 g | |
| FZIBGCDUHZBOLA-UHFFFAOYSA-N | |
| 2-fluoro-6-hydroxybenzaldehyde | |
| 2737327 | |
| ≥98.0% (GC,T) | |
| 6-Fluorosalicylaldehyde |
| 39°C | |
| MFCD01090996 | |
| 6-fluorosalicylaldehyde, 3-fluoro-2-formylphenol, benzaldehyde, 2-fluoro-6-hydroxy, 6-fluoro-2-hydroxybenzaldehyde, pubchem8482, acmc-20a0l1, 6-fluorosalicyaldehyde, ksc497m7f, 2-hydroxy-6-fluorobenzaldehyde, 2-fluoro-6-hydroxy-benzaldehyde | |
| C1=CC(=C(C(=C1)F)C=O)O | |
| 140.113 | |
| 140.11 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator