missing translation for 'onlineSavingsMsg'
Learn More

6-Fluorosalicylaldehyde 98.0+%, TCI America™

Catalog No. F09881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09881G 1 g
F09885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09881G Supplier TCI America Supplier No. F09881G

Chemical Identifiers

CAS 38226-10-7
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD01090996
InChI Key FZIBGCDUHZBOLA-UHFFFAOYSA-N
Synonym 6-fluorosalicylaldehyde, 3-fluoro-2-formylphenol, benzaldehyde, 2-fluoro-6-hydroxy, 6-fluoro-2-hydroxybenzaldehyde, pubchem8482, acmc-20a0l1, 6-fluorosalicyaldehyde, ksc497m7f, 2-hydroxy-6-fluorobenzaldehyde, 2-fluoro-6-hydroxy-benzaldehyde
PubChem CID 2737327
IUPAC Name 2-fluoro-6-hydroxybenzaldehyde
SMILES C1=CC(=C(C(=C1)F)C=O)O

Specifications

Melting Point 39°C
Quantity 1 g
Formula Weight 140.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluorosalicylaldehyde
TSCA No
Recommended Storage Refrigerator