missing translation for 'onlineSavingsMsg'
Learn More

6-Fluorosalicylaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AAL1990003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1990003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1990003 Supplier Thermo Scientific Chemicals Supplier No. L1990003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38226-10-7 | Molecular Formula: C7H5FO2 | Molecular Weight: 140.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38226-10-7
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD01090996
InChI Key FZIBGCDUHZBOLA-UHFFFAOYSA-N
Synonym 6-fluorosalicylaldehyde, 3-fluoro-2-formylphenol, benzaldehyde, 2-fluoro-6-hydroxy, 6-fluoro-2-hydroxybenzaldehyde, pubchem8482, acmc-20a0l1, 6-fluorosalicyaldehyde, ksc497m7f, 2-hydroxy-6-fluorobenzaldehyde, 2-fluoro-6-hydroxy-benzaldehyde
PubChem CID 2737327
IUPAC Name 2-fluoro-6-hydroxybenzaldehyde
SMILES C1=CC(=C(C(=C1)F)C=O)O

Specifications

Melting Point 37°C to 39°C
Quantity 1 g
Beilstein 1935289
Sensitivity Air Sensitive
Formula Weight 140.12
Percent Purity 97%
Chemical Name or Material 6-Fluorosalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only