missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroquinoline 98.0+%, TCI America™

Catalog No. F10341G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10341G 1 g
F10345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10341G Supplier TCI America Supplier No. F10341G

Chemical Identifiers

CAS 396-30-5
Molecular Formula C9H6FN
Molecular Weight (g/mol) 147.15
MDL Number MFCD01685512
InChI Key RMDCSDVIVXJELQ-UHFFFAOYSA-N
Synonym quinoline, 6-fluoro, ccris 2892, 6-fluoroquinolin, 6-fluoro-quinoline, 6-fluoro quinoline, 6-fluoro-quinolinium, 6-fluoroquinoline, acmc-209j6j
PubChem CID 196975
IUPAC Name 6-fluoroquinoline
SMILES FC1=CC=C2N=CC=CC2=C1

Specifications

Boiling Point 126°C
Quantity 1 g
Formula Weight 147.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Fluoroquinoline