missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroquinoline, 97%

Catalog No. p-7032968
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH35900MD 250 mg
AAH3590003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH35900MD Supplier Thermo Scientific Chemicals Supplier No. H35900MD
Only null left

CAS: 396-30-5 | C9H6FN | 147.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 396-30-5
Molecular Formula C9H6FN
Molecular Weight (g/mol) 147.15
MDL Number MFCD01685512
InChI Key RMDCSDVIVXJELQ-UHFFFAOYSA-N
Synonym quinoline, 6-fluoro, ccris 2892, 6-fluoroquinolin, 6-fluoro-quinoline, 6-fluoro quinoline, 6-fluoro-quinolinium, 6-fluoroquinoline, acmc-209j6j
PubChem CID 196975
IUPAC Name 6-fluoroquinoline
SMILES FC1=CC=C2N=CC=CC2=C1

Specifications

Quantity 250 mg
Formula Weight 147.15
Percent Purity 97%
Chemical Name or Material 6-Fluoroquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber VB8370020
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only